siRNA Details
| Gene Symbol: Pacs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134406.1 | NP_599233.1 | 6 | 917 | ||
| XM_006230657.2 | XP_006230719.1 | 6 | 1006 | |||
| GenBank mRNA | AF076183.1 | - | 6 | 917 | ||
| AF076184.1 | - | 6 | 917 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pacs1| Gene Symbol: Pacs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134406.1 | NP_599233.1 | 6 | 917 | ||
| XM_006230657.2 | XP_006230719.1 | 6 | 1006 | |||
| GenBank mRNA | AF076183.1 | - | 6 | 917 | ||
| AF076184.1 | - | 6 | 917 | |||