siRNA Details
| Gene Symbol: Ap1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134460.1 | NP_604455.1 | 1364 | |||
| GenBank mRNA | BC166845.1 | - | 1364 | 9029827 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap1g1| Gene Symbol: Ap1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134460.1 | NP_604455.1 | 1364 | |||
| GenBank mRNA | BC166845.1 | - | 1364 | 9029827 | ||