siRNA Details
| Gene Symbol: Atp2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134462.1 | NP_604457.1 | 10, 11 | 1009 | ||
| XM_008772595.1 | XP_008770817.1 | 4, 5 | 2989 | |||
| XM_008772596.1 | XP_008770818.1 | 4, 5 | 951 | |||
| GenBank mRNA | AF484685.1 | - | 10, 11 | 1009 | ||

Atp2c2
Genomic Map