siRNA Details
Gene Symbol: Gpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138511.1 | NP_612520.1 | 2, 3 | 551 | ||
GenBank mRNA | L20468.1 | - | 2, 3 | 551 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138511.1 | NP_612520.1 | 2, 3 | 551 | ||
GenBank mRNA | L20468.1 | - | 2, 3 | 551 |