siRNA Details
Gene Symbol: Polk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138516.1 | NP_612525.1 | 12, 13 | 1726 | ||
GenBank mRNA | BC166778.1 | - | 12, 13 | 1726 | 7934797 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Polk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138516.1 | NP_612525.1 | 12, 13 | 1726 | ||
GenBank mRNA | BC166778.1 | - | 12, 13 | 1726 | 7934797 |