siRNA Details
| Gene Symbol: Kif2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085369.1 | NP_001078838.1 | 8, 9 | 820 | ||
| NM_134472.3 | NP_608302.2 | 7, 8 | 705 | |||
| XM_006238632.2 | XP_006238694.1 | 8, 9 | 872 | |||
| XM_006238633.1 | XP_006238695.1 | 7, 8 | 636 | |||
| XM_006238634.2 | XP_006238696.1 | 7, 8 | 1168 | |||
| XM_006238635.2 | XP_006238697.1 | 8, 9 | 1045 | |||
| XM_006238636.2 | XP_006238698.1 | 8, 9 | 1041 | |||
| GenBank mRNA | BF289656.1 | - | 1, 2 | 39 | ||
| CO575506.1 | - | 7, 8 | 628 | |||

Kif2c
Genomic Map