siRNA Details
Gene Symbol: Klc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138520.1 | NP_612529.1 | 11, 12 | 1415 | ||
XM_006228378.2 | XP_006228440.1 | 11, 12 | 1568 | |||
XM_006228380.2 | XP_006228442.1 | 11, 12 | 1490 | |||
GenBank mRNA | AF166267.1 | - | 11, 12 | 1415 | ||
BC078736.1 | - | 11, 12 | 1428 | 7134584 | ||
BC097284.1 | - | 11, 12 | 1420 | 7454436 |
