siRNA Details
| Gene Symbol: A3galt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138524.2 | NP_612533.1 | 2 | 113 | ||
| GenBank mRNA | AF248543.1 | - | 2 | 112 | ||
| BC061714.1 | - | 2 | 113 | 5598151 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A3galt2| Gene Symbol: A3galt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138524.2 | NP_612533.1 | 2 | 113 | ||
| GenBank mRNA | AF248543.1 | - | 2 | 112 | ||
| BC061714.1 | - | 2 | 113 | 5598151 | ||