siRNA Details
Gene Symbol: Cdhr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138525.1 | NP_612534.1 | 15 | 2402 | ||
XM_006230508.2 | XP_006230570.1 | 15 | 2418 | |||
XM_006230509.2 | XP_006230571.1 | 14 | 2152 | |||
GenBank mRNA | AF221952.1 | - | 15 | 2402 | ||
BC098904.1 | - | 15 | 2404 | 7444666 |
