siRNA Details
| Gene Symbol: Cdhr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138525.1 | NP_612534.1 | 14 | 2123 | ||
| XM_006230508.2 | XP_006230570.1 | 14 | 2139 | |||
| XM_006230509.2 | XP_006230571.1 | 13 | 1873 | |||
| GenBank mRNA | AF221952.1 | - | 14 | 2123 | ||
| BC098904.1 | - | 14 | 2125 | 7444666 | ||

Cdhr5
Genomic Map