siRNA Details
| Gene Symbol: Nav2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138529.1 | NP_612538.1 | 9, 10 | 2257 | ||
| XM_006229244.2 | XP_006229306.1 | 10, 11 | 2769 | |||
| XM_008759312.1 | XP_008757534.1 | 9, 10 | 3279 | |||
| XM_008759313.1 | XP_008757535.1 | 9, 10 | 3279 | |||
| XM_008759314.1 | XP_008757536.1 | 9, 10 | 2380 | |||
| XM_008759315.1 | XP_008757537.1 | 4, 5 | 2597 | |||

Nav2
Genomic Map