siRNA Details
| Gene Symbol: Spon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138533.3 | NP_612542.1 | 3 | 461 | ||
| GenBank mRNA | BC078734.1 | - | 3 | 461 | 7121065 | |
| Gene Symbol: LOC100910790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770410.1 | XP_008768632.1 | 577 | |||

Spon2
Genomic Map