siRNA Details
| Gene Symbol: Grip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138535.2 | NP_612544.2 | 9 | 1083 | ||
| XM_006236895.2 | XP_006236957.1 | 9 | 2083 | |||
| XM_008763122.1 | XP_008761344.1 | 8 | 1226 | |||
| GenBank mRNA | AF072509.1 | - | 9 | 1125 | ||
| AF090113.1 | - | 1125 | ||||
| AF112182.1 | - | 9 | 987 | |||
| AF205193.1 | - | 9 | 1083 | |||

Grip2
Genomic Map