siRNA Details
| Gene Symbol: Dnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138538.1 | NP_612547.1 | 13 | 1639 | ||
| XM_006250141.2 | XP_006250203.1 | 13 | 1679 | |||
| XM_006250142.2 | XP_006250204.1 | 13 | 1679 | |||
| GenBank mRNA | AF201839.1 | - | 13 | 1639 | ||
| D14076.1 | - | 13 | 1639 | |||

Dnm3
Genomic Map