siRNA Details
| Gene Symbol: Dnase2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138539.2 | NP_612548.1 | 1, 2 | 221 | ||
| GenBank mRNA | AF178975.1 | - | 1, 2 | 222 | ||
| BC091124.1 | - | 1, 2 | 124 | 7375398 | ||
| FQ216710.1 | - | 1, 2 | 221 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnase2| Gene Symbol: Dnase2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138539.2 | NP_612548.1 | 1, 2 | 221 | ||
| GenBank mRNA | AF178975.1 | - | 1, 2 | 222 | ||
| BC091124.1 | - | 1, 2 | 124 | 7375398 | ||
| FQ216710.1 | - | 1, 2 | 221 | |||