siRNA Details
| Gene Symbol: Epcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138541.1 | NP_612550.1 | 7, 8 | 926 | ||
| GenBank mRNA | AJ001044.1 | - | 7, 8 | 981 | ||
| BC061787.1 | - | 7, 8 | 930 | |||
| BC072691.1 | - | 7, 8 | 950 | 7098920 | ||
| FQ229377.1 | - | 2, 3 | 314 | |||

Epcam
Genomic Map