siRNA Details
| Gene Symbol: Pde9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138543.1 | NP_612552.1 | 6 | 417 | ||
| XM_008772793.1 | XP_008771015.1 | 6 | 431 | |||
| XM_008772794.1 | XP_008771016.1 | 6 | 431 | |||
| XM_008772795.1 | XP_008771017.1 | 6 | 431 | |||
| GenBank mRNA | AF372654.1 | - | 6 | 417 | ||
| AY145898.1 | - | 6 | 436 | |||
| BC078733.1 | - | 6 | 358 | |||
| BC161837.1 | - | 6 | 390 | 9029236 | ||

Pde9a
Genomic Map