siRNA Details
| Gene Symbol: Serpina12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138825.2 | NP_620180.2 | 3 | 964 | ||
| GenBank mRNA | AF245398.1 | - | 3 | 964 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpina12| Gene Symbol: Serpina12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138825.2 | NP_620180.2 | 3 | 964 | ||
| GenBank mRNA | AF245398.1 | - | 3 | 964 | ||