siRNA Details
| Gene Symbol: Syt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138835.1 | NP_620190.1 | 7 | 1272 | ||
| XM_008760071.1 | XP_008758293.1 | 7 | 1200 | |||
| XM_008760072.1 | XP_008758294.1 | 8 | 1246 | |||
| GenBank mRNA | U71294.1 | - | 7 | 1272 | ||

Syt12
Genomic Map