siRNA Details
| Gene Symbol: Syngap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113409.2 | NP_001106880.2 | 7 | 1451 | ||
| NM_181092.4 | NP_851606.3 | 9 | 1514 | |||
| GenBank mRNA | AF048976.1 | - | 9 | 1522 | ||
| AF050183.2 | - | 5 | 1203 | |||
| AF058789.2 | - | 9 | 1401 | |||
| AF058790.1 | - | 7 | 1269 | |||

Syngap1
Genomic Map