siRNA Details
| Gene Symbol: Sftpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138842.1 | NP_620197.1 | 4 | 279 | ||
| XM_006236657.2 | XP_006236719.1 | 4 | 314 | |||
| GenBank mRNA | BC072466.1 | - | 4 | 278 | 7100971 | |
| X14778.1 | - | 4 | 279 | |||

Sftpb
Genomic Map