siRNA Details
Gene Symbol: Mpst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138843.1 | NP_620198.1 | 1 | 713 | ||
GenBank mRNA | BC086575.1 | - | 1 | 528 | 7306783 | |
D50564.1 | - | 1 | 713 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mpst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138843.1 | NP_620198.1 | 1 | 713 | ||
GenBank mRNA | BC086575.1 | - | 1 | 528 | 7306783 | |
D50564.1 | - | 1 | 713 |