siRNA Details
| Gene Symbol: Ubxn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138853.2 | NP_620208.1 | 9 | 765 | ||
| XM_006239000.1 | XP_006239062.1 | 9 | 827 | |||
| XM_006239002.1 | XP_006239064.1 | 7 | 701 | |||
| XM_006239004.1 | XP_006239066.1 | 7 | 681 | |||
| XM_006239005.2 | XP_006239067.1 | 6 | 1035 | |||
| XM_008764131.1 | XP_008762353.1 | 7 | 833 | |||
| XM_008764132.1 | XP_008762354.1 | 7 | 874 | |||
| GenBank mRNA | AB072920.1 | - | 8 | 693 | ||
| BC078730.1 | - | 9 | 765 | 7134156 | ||

Ubxn11
Genomic Map