siRNA Details
| Gene Symbol: Slc38a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138854.1 | NP_620209.1 | 14 | 1309 | ||
| XM_008773068.1 | XP_008771290.1 | 14 | 1241 | |||
| GenBank mRNA | AF276870.1 | - | 14 | 1309 | ||
| BC128725.1 | - | 14 | 1241 | 8362014 | ||

Slc38a5
Genomic Map