siRNA Details
| Gene Symbol: Slc38a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138854.1 | NP_620209.1 | 14 | 1253 | ||
| GenBank mRNA | AF276870.1 | - | 14 | 1253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc38a5| Gene Symbol: Slc38a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138854.1 | NP_620209.1 | 14 | 1253 | ||
| GenBank mRNA | AF276870.1 | - | 14 | 1253 | ||