siRNA Details
| Gene Symbol: Spdya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138855.1 | NP_620210.1 | 6 | 653 | ||
| XM_006239766.1 | XP_006239828.1 | 6 | 887 | |||
| XM_006239767.2 | XP_006239829.1 | 6 | 851 | |||
| XM_006239768.2 | XP_006239830.1 | 6 | 718 | |||
| XM_008764502.1 | XP_008762724.1 | 6 | 1553 | |||
| GenBank mRNA | AF492385.1 | - | 6 | 653 | ||
| BC078729.1 | - | 5 | 576 | 7134092 | ||

Spdya
Genomic Map