siRNA Details
| Gene Symbol: Kat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005872.1 | NP_001005872.1 | 6 | 812 | ||
| GenBank mRNA | AF333984.1 | - | 4 | 465 | ||
| BC083879.1 | - | 6 | 812 | 7130366 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kat5| Gene Symbol: Kat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005872.1 | NP_001005872.1 | 6 | 812 | ||
| GenBank mRNA | AF333984.1 | - | 4 | 465 | ||
| BC083879.1 | - | 6 | 812 | 7130366 | ||