siRNA Details
| Gene Symbol: Spata7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138862.3 | NP_620217.2 | 6 | 548 | ||
| XM_006240413.2 | XP_006240475.1 | 5 | 531 | |||
| XM_006240414.2 | XP_006240476.1 | 6 | 623 | |||
| XM_006240415.2 | XP_006240477.1 | 4 | 456 | |||
| XM_008764737.1 | XP_008762959.1 | 5 | 502 | |||
| GenBank mRNA | BC078728.1 | - | 6 | 538 | 7114267 | |
| CK470849.1 | - | 2 | 212 | |||
| CK600391.1 | - | 5 | 520 | |||

Spata7
Genomic Map