siRNA Details
| Gene Symbol: Cand2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181362.1 | NP_852027.1 | 8 | 1229 | ||
| XM_006237004.2 | XP_006237066.1 | 10 | 1346 | |||
| GenBank mRNA | AB029324.1 | - | 8 | 1229 | ||
| AB029341.1 | - | 10 | 1343 | |||
| AB029342.1 | - | 7 | 1368 | |||

Cand2
Genomic Map