siRNA Details
| Gene Symbol: Ptgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138863.2 | NP_620218.1 | 8, 9 | 806 | ||
| GenBank mRNA | BC089775.1 | - | 8, 9 | 806 | 7307365 | |
| FQ218841.1 | - | 8, 9 | 825 | |||
| FQ220898.1 | - | 8, 9 | 827 | |||
| FQ229147.1 | - | 8, 9 | 822 | |||
| FQ229679.1 | - | 8, 9 | 822 | |||
| U66322.2 | - | 7, 8 | 771 | |||

Ptgr1
Genomic Map