siRNA Details
| Gene Symbol: Eif2b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138866.2 | NP_620221.2 | 5, 6 | 788 | ||
| GenBank mRNA | BC085698.1 | - | 5, 6 | 788 | 7193458 | |
| U19516.1 | - | 5, 6 | 786 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2b5| Gene Symbol: Eif2b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138866.2 | NP_620221.2 | 5, 6 | 788 | ||
| GenBank mRNA | BC085698.1 | - | 5, 6 | 788 | 7193458 | |
| U19516.1 | - | 5, 6 | 786 | |||