siRNA Details
| Gene Symbol: Akr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138884.1 | NP_620239.1 | 1, 2 | 138 | ||
| GenBank mRNA | D17309.1 | - | 1, 2 | 138 | ||
| S80431.1 | - | 1, 2 | 110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akr1d1| Gene Symbol: Akr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138884.1 | NP_620239.1 | 1, 2 | 138 | ||
| GenBank mRNA | D17309.1 | - | 1, 2 | 138 | ||
| S80431.1 | - | 1, 2 | 110 | |||