siRNA Details
Gene Symbol: Cdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138889.2 | NP_620244.1 | 9 | 1206 | ||
GenBank mRNA | AF494095.1 | - | 9 | 1203 | ||
BC085699.1 | - | 9 | 1206 | 7106186 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138889.2 | NP_620244.1 | 9 | 1206 | ||
GenBank mRNA | AF494095.1 | - | 9 | 1203 | ||
BC085699.1 | - | 9 | 1206 | 7106186 |