siRNA Details
| Gene Symbol: Ehd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138890.2 | NP_620245.2 | 6 | 1624 | ||
| GenBank mRNA | AF494093.2 | - | 6 | 1624 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ehd3| Gene Symbol: Ehd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138890.2 | NP_620245.2 | 6 | 1624 | ||
| GenBank mRNA | AF494093.2 | - | 6 | 1624 | ||