siRNA Details
| Gene Symbol: Gpr149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138891.1 | NP_620246.1 | 4 | 2676 | ||
| GenBank mRNA | AY030276.1 | - | 4 | 2676 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr149| Gene Symbol: Gpr149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138891.1 | NP_620246.1 | 4 | 2676 | ||
| GenBank mRNA | AY030276.1 | - | 4 | 2676 | ||
Set Membership: |
|