siRNA Details
| Gene Symbol: Myo16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138893.1 | NP_620248.1 | 31 | 4093 | ||
| XM_008771384.1 | XP_008769606.1 | 32 | 5067 | |||
| GenBank mRNA | AY004215.1 | - | 31 | 4093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo16| Gene Symbol: Myo16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138893.1 | NP_620248.1 | 31 | 4093 | ||
| XM_008771384.1 | XP_008769606.1 | 32 | 5067 | |||
| GenBank mRNA | AY004215.1 | - | 31 | 4093 | ||