siRNA Details
| Gene Symbol: Myo16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138893.1 | NP_620248.1 | 17 | 2106 | ||
| XM_008771384.1 | XP_008769606.1 | 18 | 3080 | |||
| GenBank mRNA | AF209114.1 | - | 17 | 2106 | ||
| AY004215.1 | - | 17 | 2106 | |||

Myo16
Genomic Map