siRNA Details
| Gene Symbol: Pja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277278.1 | NP_001264207.1 | 7 | 1835 | ||
| NM_138896.2 | NP_620251.1 | 8 | 2021 | |||
| GenBank mRNA | BC074015.1 | - | 8 | 1946 | 7105756 | |
| D32249.1 | - | 8 | 1957 | |||

Pja2
Genomic Map