siRNA Details
| Gene Symbol: Ppp1r3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138912.2 | NP_620267.2 | 2 | 1035 | ||
| XM_006253215.2 | XP_006253277.1 | 2 | 1601 | |||
| XM_006253216.2 | XP_006253278.1 | 2 | 922 | |||
| GenBank mRNA | BC072514.1 | - | 2 | 1035 | 7104413 | |
| Y18208.1 | - | 2 | 898 | |||

Ppp1r3b
Genomic Map