siRNA Details
| Gene Symbol: Gmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013036.1 | NP_001013054.1 | 3 | 250 | ||
| XM_006251925.2 | XP_006251987.1 | 3 | 311 | |||
| XM_006251926.2 | XP_006251988.1 | 3 | 311 | |||
| XM_006251927.2 | XP_006251989.1 | 3 | 332 | |||
| GenBank mRNA | AF350372.1 | - | 3 | 206 | ||
| BC089848.1 | - | 3 | 250 | 7374684 | ||
| FQ211507.1 | - | 3 | 272 | |||

Gmpr2
Genomic Map