siRNA Details
| Gene Symbol: Mfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138976.1 | NP_620432.1 | 4 | 512 | ||
| XM_008760904.1 | XP_008759126.1 | 6 | 759 | |||
| GenBank mRNA | AB084166.1 | - | 4 | 512 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfn1| Gene Symbol: Mfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138976.1 | NP_620432.1 | 4 | 512 | ||
| XM_008760904.1 | XP_008759126.1 | 6 | 759 | |||
| GenBank mRNA | AB084166.1 | - | 4 | 512 | ||