siRNA Details
| Gene Symbol: Rin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139038.1 | NP_620607.1 | 10 | 2215 | ||
| XM_008760073.1 | XP_008758295.1 | 2641 | ||||
| GenBank mRNA | U80076.1 | - | 10 | 2215 | ||
| Gene Symbol: LOC100911794 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749037.3 | XP_003749085.1 | 10 | 2497 | ||
| XM_008760195.1 | XP_008758417.1 | 9 | 2641 | |||

Rin1
Genomic Map