siRNA Details
| Gene Symbol: Muc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768788.1 | XP_008767010.1 | 7 | 1184 | ||
| GenBank mRNA | U89744.1 | - | 6 | 1146 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Muc13| Gene Symbol: Muc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768788.1 | XP_008767010.1 | 7 | 1184 | ||
| GenBank mRNA | U89744.1 | - | 6 | 1146 | ||