siRNA Details
| Gene Symbol: A2m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012488.2 | NP_036620.2 | 12 | 1479 | ||
| GenBank mRNA | AY921651.1 | - | 12 | 1438 | ||
| BC098922.1 | - | 12 | 1479 | 7458629 | ||
| FQ223246.1 | - | 12 | 1501 | |||
| FQ228339.1 | - | 12 | 1678 | |||
| J02635.1 | - | 12 | 1501 | |||
| M11793.1 | - | 2 | 181 | |||
| Gene Symbol: LOC100911545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749828.2 | XP_003749876.1 | 1785 | |||

A2m
Genomic Map