siRNA Details
| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016986.2 | NP_058682.2 | 4 | 361 | ||
| GenBank mRNA | CK359511.1 | - | 4 | 276 | ||
| J02791.1 | - | 4 | 276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acadm| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016986.2 | NP_058682.2 | 4 | 361 | ||
| GenBank mRNA | CK359511.1 | - | 4 | 276 | ||
| J02791.1 | - | 4 | 276 | |||