siRNA Details
| Gene Symbol: Acp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016988.2 | NP_058684.2 | 9, 10 | 1008 | ||
| XM_006234500.1 | XP_006234562.1 | 10, 11 | 1930 | |||
| GenBank mRNA | BC081823.1 | - | 9, 10 | 979 | 7107570 | |
| M27893.1 | - | 9, 10 | 977 | |||

Acp2
Genomic Map