siRNA Details
| Gene Symbol: Afp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012493.2 | NP_036625.2 | 6 | 676 | ||
| GenBank mRNA | BC097344.1 | - | 6 | 659 | 7459964 | |
| DY309525.1 | - | 6 | 676 | |||
| V01254.1 | - | 3 | 389 | |||
| X02361.1 | - | 6 | 680 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Afp| Gene Symbol: Afp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012493.2 | NP_036625.2 | 6 | 676 | ||
| GenBank mRNA | BC097344.1 | - | 6 | 659 | 7459964 | |
| DY309525.1 | - | 6 | 676 | |||
| V01254.1 | - | 3 | 389 | |||
| X02361.1 | - | 6 | 680 | |||