siRNA Details
| Gene Symbol: Afp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012493.2 | NP_036625.2 | 9 | 1126 | ||
| GenBank mRNA | BC097344.1 | - | 9 | 1109 | 7459964 | |
| DY317147.1 | - | 4 | 457 | |||
| FQ210921.1 | - | 2 | 154 | |||
| V01254.1 | - | 6 | 839 | |||
| X02361.1 | - | 9 | 1130 | |||

Afp
Genomic Map