siRNA Details
| Gene Symbol: Aldoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012497.1 | NP_036629.1 | 2 | 151 | ||
| GenBank mRNA | BC059154.1 | - | 2 | 126 | ||
| BC099749.1 | - | 2 | 137 | 5599032 | ||
| FQ213506.1 | - | 2 | 154 | |||
| FQ214420.1 | - | 2 | 177 | |||
| FQ228893.1 | - | 2 | 151 | |||
| X06984.1 | - | 2 | 151 | |||

Aldoc
Genomic Map