siRNA Details
Gene Symbol: Apeh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012500.1 | NP_036632.1 | 18, 19 | 1686 | ||
XM_008766476.1 | XP_008764698.1 | 17, 18 | 1790 | |||
GenBank mRNA | BC166605.1 | - | 18, 19 | 1692 | 7937766 | |
J04733.1 | - | 18, 19 | 1686 |
